Order Entry
VWR®, Multi-Parameter Refractive Index and Density Certified Reference Material At 15, 20 and 25 °C
undefined
VWR®, Multi-Parameter Refractive Index and Density Certified Reference Material At 15, 20 and 25 °C
  •  
    Density
    Nominal refractive index at 25 °C
    Reference temperature
    Pack type
    Pk
  • Refractive Index & Density Certified Reference Material
    Refractive Index & Density Certified Reference Material
    99173.030
    0.99705 g/ml
    1.33249 
    25 °C
    Glass bottle
    30 mL
    1 * 30 mL
    1 * 30 mL - 9 920,00 Kč
    9 920,00 Kč
    1 * 30 mL
       9 920,00 Kč
  • Refractive Index & Density Certified Reference Material
    Refractive Index & Density Certified Reference Material
    99174.030
    0.74520 g/ml
    1.41959  
    25 °C
    Glass bottle
    30 mL
    1 * 30 mL
    1 * 30 mL - 9 920,00 Kč
    9 920,00 Kč
    1 * 30 mL
       9 920,00 Kč
  • Refractive Index & Density Certified Reference Material
    Refractive Index & Density Certified Reference Material
    99175.030
    1.24430 g/ml
    1.54403 
    25 °C
    Glass bottle
    30 mL
    1 * 30 mL
    1 * 30 mL - 9 920,00 Kč
    9 920,00 Kč
    1 * 30 mL
       9 920,00 Kč
  • Refractive Index & Density Certified Reference Material
    Refractive Index & Density Certified Reference Material
    99176.030
    1.47751 g/ml
    1.65555
    25 °C
    Glass bottle
    30 mL
    1 * 30 mL
    1 * 30 mL - 9 920,00 Kč
    9 920,00 Kč
    1 * 30 mL
       9 920,00 Kč