J.T.Baker® BAKERBOND® PROchievA™, Affinity Chromatography Columns, Recombinant Protein A Resin
: Avantor
- Catalog No:
- C789-25
- C789-07
- C789-11
- C789-18
- Binding capacity:
- FIO (Target > 65 mg/ml)
- FIO (Target > 65 mg/ml)
- FIO (Target > 65 mg/ml)
- FIO (Target > 65 mg/ml)
- Storage:
- +2...+8 °C
- +2...+8 °C
- +2...+8 °C
- +2...+8 °C
Avantor J.T.Baker BAKERBOND PROchievA column packing guidelines Technical Note Avantor J.T.Baker BAKERBOND PROchievA pre-packed column guideline Avantor J.T.Baker Binding affinity of PROchievA resin to IgG-Technical Note Avantor J.T.Baker Specific binding to Fc domain of BAKERBOND PROchievA resin-Technical Note
- CapacityPk
- BAKERBOND® PROchievA™ protein A resin, lab column, box of 1BAKERBOND® PROchievA™ protein A resin, lab column, box of 11 ml11 * 1 KS6 190,00 Kč
Accessories
Specifications
Environmentally PreferableBinding capacityFIO (Target > 65 mg/ml)Storage+2...+8 °C - BAKERBOND® PROchievA™ protein A resin, lab column, box of 5BAKERBOND® PROchievA™ protein A resin, lab column, box of 51 ml51 * 5 KS22 380,00 Kč
Accessories
Specifications
Environmentally PreferableBinding capacityFIO (Target > 65 mg/ml)Storage+2...+8 °C - BAKERBOND® PROchievA™ protein A resin, lab column, box of 1BAKERBOND® PROchievA™ protein A resin, lab column, box of 15 ml11 * 1 KS23 940,00 Kč
Accessories
Specifications
Environmentally PreferableBinding capacityFIO (Target > 65 mg/ml)Storage+2...+8 °C - BAKERBOND® PROchievA™ protein A resin, lab column, box of 5BAKERBOND® PROchievA™ protein A resin, lab column, box of 55 ml51 * 5 KS93 770,00 Kč
Accessories
Specifications
Environmentally PreferableBinding capacityFIO (Target > 65 mg/ml)Storage+2...+8 °C