Shed some light on inspections by supplementing inspection tags with the inspection timer, and get an inspection status at a glance.
- Unlike tags, the bright colour LED lights provide an inspection status from a distance and break through the grime of any facility
- Review and reset; after completing an inspection, reset the timer so it is ready for the next inspection
- Perfect for facilities that have internal audit and inspection schedules
- Sturdy ABS plastic construction
- IP 65 rated timers will survive harsh industrial environments, including exposure to water, dust, extreme temperatures, and more
Safety inspection timers are ideal for use in facilities that have multiple eyewash stations, emergency showers, emergency exit doors, exit signs, fire extinguishers, or first aid kits.
Certifikace: Ingress Protection Rating IP 65.