Tisk…
Protective screen, transparent
Dodavatel: Gett-Group
GETTSC27009EA 2000 CZK
GETTSC27009 GETTSC27010
Protective screen, transparent
Protective screens are designed to protect the user and their environment from droplet infections that can be transmitted through speaking, coughing, or sneezing. Equipped with two self-supporting feet, the screen is stable on flat surfaces such as counters or tables.
  • Shatterproof polycarbonate material
  • Suitable for indoor use
  • Self-supporting due to plug-in feet
  • Holes available for hanging option
  • Transparent material is not tinted
  • Can be cleaned with ordinary disinfectant
Screens can be cleaned with a lint-free cloth, water, and 70% isopropyl alcohol. Do not use scratching cloths or sponges for cleaning, as the surface is not scratch-resistant.