Tisk…
Signs, COVID-19 floor signs, no text
Dodavatel: Brady
116-0934EA 705 CZK
116-0934 116-0933 116-0936 116-0935 116-0927 116-0937 116-0929 116-0928 116-0930 116-0932 116-0931
Signs, COVID-19 floor signs, no text
Značení bezpečnostní
Signs are made of surface printed anti-skid laminated white polyester with a permanent acrylic pressure-sensitive rubber adhesive. They are overlaminated with a clear anti-slip vinyl film.
  • Anti-slip safety sign
  • Ideal for floormarking applications in stairs, hallways, and offices
Thickness: 0,254 mm.
Certifikace: Tested according DIN 51130:2014 with a slip resistance classification of R9.