Tisk…
Hledali jste: Krémy pro pokožku
24  výsledků nalezeno
SearchResultCount:"24"
Sort Results
Zobrazit seznam Rychlý náhled (novinka)
Ohodnoťte výsledky hledání
Dodavatel: Deb-STOKO
Popis: Softcream (O/W emulsion) with moisturising effect for the effective care of normal skin. Suitable for all over body use and for use under latex and ni...
Dodavatel: Deb-STOKO
Popis: Soft cream (O/W emulsion) with moisturising effect for the effective care of normal skin. Suitable for all over body use. Suitable for people prone to...
Dodavatel: Deb-STOKO
Popis: Aqua PURE is a barrier cream (W/O/W emulsion) that protects the skin when working with aqueous or water-miscible industrial substances and performing ...
Katalogové číslo: (115-2250)
Dodavatel: Deb-STOKO
Popis: Triple action cream protects against water and oil based substances, plus helps improve grip and comfort under gloves. Perfume-free and dye-free.
Měrná jednotka: 1 * 9 KS
Dodavatel: Deb-STOKO
Popis: Stokolan® Intense is a nurturing, regenerative and barrier active creamy lotion containing grapeseed oil, ceramid complex, allantoin, ginkgo biloba, u...
Dodavatel: Deb-STOKO
Popis: Versatile skin protection product (O/W emulsion) for sensitive skin exposed to changing and non specific stresses. Can also be used under gloves. The ...
Dodavatel: Deb-STOKO
Popis: Speciální krém používaný před prací k ochraně proti vzdorným, ve vodě nerozpustným nečistotám jako jsou oleje, barvy, grafit, kovový prach, adhezivní ...
Akce
Katalogové číslo: (115-1743)
Dodavatel: Deb-STOKO
Popis: White soft cream (O/W emulsion) highly suitable for sensitive skin and for facial care. Dermatologically confirmed.
Měrná jednotka: 1 * 9 KS
Katalogové číslo: (EVON99036430)
Dodavatel: Deb-STOKO
Popis: This intensive care (W/O) cream contains a ceramide complex and grapeseed oil. It is ideal for dry and sensitive skin and regenerates and deeply nurtu...
Měrná jednotka: 1 * 9 KS
Dodavatel: Deb-STOKO
Popis: Perfume-free, water resistant 5-star rated, factor 50 sunscreen prolongs the skin's natural protection time against UVA, UVB and UVC exposure. For use...
Dodavatel: Deb-STOKO
Popis: Universal pre-work cream to help protect the skin against water and non water-based workplace contaminants, such as oils and solvents.
Dodavatel: Deb-STOKO
Popis: Stokolan® classic is a concentrated moisturising and conditioning cream that helps to restore moisture in normal and dry skin. Ideal for use in indust...
Katalogové číslo: (115-2354)
Dodavatel: Deb-STOKO
Popis: This foot spray helps to protect feet by preventing excessive perspiration, for instance when wearing closed in footwear, such as safety shoes, rubber...
Měrná jednotka: 1 * 25 KS
Katalogové číslo: (115-1339)
Dodavatel: Deb-STOKO
Popis: A concentrated moisturising and conditioning cream to help restore moisture to dry and stressed skin. Suitable for all over body use and for use under...
Měrná jednotka: 1 * 9 KS
Katalogové číslo: (DIVY101100799)
Dodavatel: Diversey
Popis: Cleans and disinfects hands in one easy step. Based on lactic acid, an organic based ingredient from Natural sources that inhibits the growth of micro...
Měrná jednotka: 1 * 4 Bottle
Udržitelná balení
Udržitelné materiály
Dodavatel: Diversey
Popis: Gentle and effective hand wash with a 100% natural perfume. Cleans hands leaving them hygienically fresh.
Cena na vyžádání
Omezené množství produktu na skladě. Zboží může být k dispozici v jiném skladě poblíž vašeho sídla. Přesvědčte se, že jste přihlášení na stránky, abyste mohli vidět dostupné položky na skladě. Pokud je stále zobrazeno call a potřebujete asistenci, volejte na číslo 321 570 321.
Omezené množství produktu na skladě. Zboží může být k dispozici v jiném skladě poblíž vašeho sídla. Přesvědčte se, že jste přihlášení na stránky, abyste mohli vidět dostupné položky na skladě. Pokud je stále zobrazeno call a potřebujete asistenci, volejte na číslo 321 570 321.
Daná chemikálie je regulována dle platné legislativy a bude vyžadováno vyplnění formuláře. Jeho včasným vyplněním uspíšíte dodání produktu.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Tento produkt je zablokován vaší organizací. Kontaktujte své nákupní oddělení pro více informací.
Původní produkt již není k dispozici. Zobrazen je náhradní produkt.
Tento produkt již není k dispozici. Podobné produkty můžete vyhledat pomocí VWR katalogových čísel nebo čísel dodavatele uvedených výše. Pokud potřebujete další asistenci, zavolejte na telefonní číslo zákaznického servisu VWR 321 570 321.
1 - 16 of 24
no targeter for Bottom