Tisk…
Hledali jste: Rohože proti únavě
20  výsledků nalezeno
SearchResultCount:"20"
Sort Results
Zobrazit seznam Rychlý náhled (novinka)
Ohodnoťte výsledky hledání
Dodavatel: VWR Collection
Popis: Ergonomické laboratorní podložky nabízí pohodlí polyuretanu a jsou vhodné pro čistá prostředí, která podléhají normě ISO třída 5 (třída 100). Jsou k d...
Dodavatel: VWR Collection
Popis: Podložky jsou vyrobeny z polyuretanu o nízké hustotě a poskytují ergonomický a stimulující povrch. Bublinkové podložky (Bubble) jsou určeny pro suchá ...
Katalogové číslo: (115-1060)
Dodavatel: VWR Collection
Popis: Anti-fatigue mats, VWR®, Pro čisté prostory: +, D׊: 600×900 mm
Měrná jednotka: 1 * 1 KS
Dodavatel: JUSTRITE MANUFACTURING CO
Popis: The Skystep™ ESD static dissipative mat is a stand-alone rubber mat designed for individual work stations. Ergonomically engineered, the SkyStep™ ESD ...
Katalogové číslo: (JUSU549S3124OB)
Dodavatel: JUSTRITE MANUFACTURING CO
Popis: High quality anti-fatigue matting for use in areas where there can be frequent oil spilling.
Měrná jednotka: 1 * 1 KS
New Product
Katalogové číslo: (JUSU470S2436GY)
Dodavatel: JUSTRITE MANUFACTURING CO
Popis: Marble Sof-Tyle™ is an anti-fatigue floor mat provides comfort and ergonomic support and complements any décor with a stylish smooth marble-pattern de...
Měrná jednotka: 1 * 1 KS
New Product
Katalogové číslo: (MMMA3270E930)
Dodavatel: 3M
Popis: Safety Walk™ Light duty cushion matting 3270E is made from continuous coiled vinyl filaments that provide comfort and support across the bottom of the...
Měrná jednotka: 1 * 1 KS
Katalogové číslo: (JUSU562S0035BL)
Dodavatel: JUSTRITE MANUFACTURING CO
Popis: Sanitop® mats are made from a durable rubber compound designed to withstand harsh chemicals while creating a slip resistant surface.
Měrná jednotka: 1 * 1 KS
New Product
Dodavatel: ERGOMAT
Popis: The Infinity Bubble mat has the comfort of a polyurethane mat and easy clean properties due to its closed-cell and non-porous top surface. It is UV an...
Dodavatel: ERGOMAT
Popis: Ergomat® Hygiene is made from pure nitrile rubber and is designed to increase comfort while reducing trip hazards, body pain, quality issues and sick ...
Dodavatel: ERGOMAT
Popis: The anti-static Ergomat® AFS Complete is made of pure polyurethane and is designed to increase comfort while reducing trip hazards and body pain. This...
Dodavatel: ERGOMAT
Popis: The Ergomat® Nitril and Nitril Smooth are made from pure nitrile rubber and is designed to increase comfort while reducing trip hazards, body pain, qu...
Dodavatel: ERGOMAT
Popis: Ergomat® Nitro with anti-slip cone profile is made from pure nitrile rubber and is designed to increase comfort while reducing trip hazards, body pain...
Dodavatel: ERGOMAT
Popis: The Ergomat Softline is made from nitrile rubber and is designed to increase comfort while reducing tripping hazards, body aches, quality issues, and ...
Dodavatel: ERGOMAT
Popis: The Ergomat® Nitril ESD Conductive is made from pure nitrile rubber and is designed to increase comfort while reducing trip hazards, body pain, qualit...
Dodavatel: ERGOMAT
Popis: The anti-static Ergomat® AB Classic is made of pure polyurethane and is designed to increase comfort while reducing trip hazards and body pain. It is ...
Cena na vyžádání
Omezené množství produktu na skladě. Zboží může být k dispozici v jiném skladě poblíž vašeho sídla. Přesvědčte se, že jste přihlášení na stránky, abyste mohli vidět dostupné položky na skladě. Pokud je stále zobrazeno call a potřebujete asistenci, volejte na číslo 321 570 321.
Omezené množství produktu na skladě. Zboží může být k dispozici v jiném skladě poblíž vašeho sídla. Přesvědčte se, že jste přihlášení na stránky, abyste mohli vidět dostupné položky na skladě. Pokud je stále zobrazeno call a potřebujete asistenci, volejte na číslo 321 570 321.
Daná chemikálie je regulována dle platné legislativy a bude vyžadováno vyplnění formuláře. Jeho včasným vyplněním uspíšíte dodání produktu.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Tento produkt je zablokován vaší organizací. Kontaktujte své nákupní oddělení pro více informací.
Původní produkt již není k dispozici. Zobrazen je náhradní produkt.
Tento produkt již není k dispozici. Podobné produkty můžete vyhledat pomocí VWR katalogových čísel nebo čísel dodavatele uvedených výše. Pokud potřebujete další asistenci, zavolejte na telefonní číslo zákaznického servisu VWR 321 570 321.
1 - 16 of 20
no targeter for Bottom