Tisk…
Hledali jste: Lázně s odnímatelným oběhovým čerpadlem
15  výsledků nalezeno
SearchResultCount:"15"
Sort Results
Zobrazit seznam Rychlý náhled (novinka)
Ohodnoťte výsledky hledání
Dodavatel: JULABO GmbH
Popis: Výkonný a robustní termostat/lázeň CORIO™ poskytuje přesnou teplotní regulaci, absolutní preciznost a široké rozpětí pracovních teplot. Imerzní termos...
Dodavatel: JULABO GmbH
Popis: The powerful and robust CORIO™ laboratory circulators provide the exact temperature, absolute precision and a wide working temperature range. The CORI...
Akce
Katalogové číslo: (LAUDL001098)
Dodavatel: LAUDA
Popis: ECO units with transparent plastic baths provide visibility in all cases where test samples need to be observed during thermostating. The thermostats ...
Měrná jednotka: 1 * 1 KS
Dodavatel: IKA
Popis: The HBC circulating baths have a high-quality stainless steel bath, an extended temperature range for tempering fluids in open and covered water baths...
Dodavatel: HUBER
Popis: These circulators with insulated refrigeration baths are designed for direct temperature control from −20 to +200 °C. The units are suitable for ...
Dodavatel: HUBER
Popis: Heating circulation bath thermostats with either polycarbonate or stainless steel baths, which are simple to use and are suitable for a variety of app...
Dodavatel: HUBER
Popis: Ces cryothermostats à circulation sont parfaits pour le contrôle direct de la température de −30 à +200 °C.
Dodavatel: HUBER
Popis: Heating circulation bath thermostats with polycarbonate or stainless steel baths, which are simple to use and are suitable for a variety of applicatio...
Dodavatel: IKA
Popis: The ICC compact immersion circulators allow flexible coupling to water baths of different volumes. They are designed for use with liquids up to temper...
Dodavatel: HUBER
Popis: Circulating bath thermostats with baths made from stainless steel are simple to use and suitable for many applications such as: sample temperature con...
Dodavatel: IKA
Popis: The ICC compact immersion circulators allow flexible coupling to water baths of different volumes. They are designed for use with liquids up to temper...
Dodavatel: THERMO ELECTRON LED GMBH
Popis: These heated bath circulators maintains the ambient temperature. The SAHARA series of heated bath circulators are available with a wide variety of wor...
Dodavatel: NICKEL ELECTRO
Popis: These circulating water baths are available in two ranges for circulating just water up to 99 °C or oil up to 130 °C. Operating temperatures below amb...
Dodavatel: NICKEL ELECTRO
Popis: These thermostatically controlled analogue stirred baths have a corrosion resistant stainless steel tank and durable outer case. Immersed components a...
Dodavatel: THERMO ELECTRON LED GMBH
Popis: The SAHARA series are suitable for internal or external circulation and have a choice of eight different controllers that are suitable for various dif...
Cena na vyžádání
Omezené množství produktu na skladě. Zboží může být k dispozici v jiném skladě poblíž vašeho sídla. Přesvědčte se, že jste přihlášení na stránky, abyste mohli vidět dostupné položky na skladě. Pokud je stále zobrazeno call a potřebujete asistenci, volejte na číslo 321 570 321.
Omezené množství produktu na skladě. Zboží může být k dispozici v jiném skladě poblíž vašeho sídla. Přesvědčte se, že jste přihlášení na stránky, abyste mohli vidět dostupné položky na skladě. Pokud je stále zobrazeno call a potřebujete asistenci, volejte na číslo 321 570 321.
Daná chemikálie je regulována dle platné legislativy a bude vyžadováno vyplnění formuláře. Jeho včasným vyplněním uspíšíte dodání produktu.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Tento produkt je zablokován vaší organizací. Kontaktujte své nákupní oddělení pro více informací.
Původní produkt již není k dispozici. Zobrazen je náhradní produkt.
Tento produkt již není k dispozici. Podobné produkty můžete vyhledat pomocí VWR katalogových čísel nebo čísel dodavatele uvedených výše. Pokud potřebujete další asistenci, zavolejte na telefonní číslo zákaznického servisu VWR 321 570 321.
1 - 15 of 15
no targeter for Bottom