Tisk…
Soap solution according to Boutron-Boudet
Dodavatel: VWR Chemicals
31700.293EA 1730 CZK
31700.293_T 31700.293
Soap solution according to Boutron-Boudet
Kity pro environmentální testy
Solution for water hardness testing.
0,104 ml corresponds to 1 hydrotimetric degree for a 40 ml water sample