Hexacarbonylmolybdenum ≥98%
|
Nebezpečí
|
13057.14EA
6080
CZK
13057.14
13057.22
13057.88
Hexacarbonylmolybdenum ≥98%
Hexacarbonylmolybdenum
Vzorec:
C₆MoO₆ MW: 264 g/mol Bod varu: 155 °C (1013 hPa) Bod tání: ∼ 150 °C (rozkl.) Hustota: 1,96 Storage Temperature: Ambient |
MDL Number:
MFCD00003466 CAS číslo: 13939-06-5 EINECS: 237-713-3 UN: 3466 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...