Hexacarbonylmolybdenum for synthesis, Sigma-Aldrich®
|
|
M-Clarity™ quality level = MQ200
8.00396.0010EA
6960
CZK
8.00396.0010
8.00396.9999
8.00396.0025
Hexacarbonylmolybdenum for synthesis, Sigma-Aldrich®
Hexacarbonylmolybdenum
Vzorec:
C₆MoO₆ MW: 264 g/mol Bod varu: 155 °C (1013 hPa) Hustota: 1,96 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00003466 CAS číslo: 13939-06-5 EINECS: 237-713-3 ADR: 6.1,III |
Výsledky specifikačního testu
Solubility | (20 °C) insoluble |
Melting Point | 147 - 152 °C (decomposition) |
Molar Mass | 264 g/mol |
Boiling Point | 156.4 °C |
Vapor Pressure | 0.13 hPa (20 °C) |
Density | 1.96 g/cm3 (20 °C) |
Synonyms | Molybdenum carbonyl |
Hill Formula | C6MoO6 |
Chemical Formula | Mo(CO)6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...