Tisk…

Hexacarbonylmolybdenum, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIAL69875-25GEA 5600 CZK
SIAL69875-25G SIAL69875-100G
Hexacarbonylmolybdenum, Sigma-Aldrich®
Hexacarbonylmolybdenum
Vzorec: C₆MoO₆
MW: 264 g/mol
Bod varu: 155 °C (1013 hPa)
Hustota: 1,96 g/cm³ (20 °C)
MDL Number: MFCD00003466
CAS číslo: 13939-06-5
EINECS: 237-713-3
UN: 3466
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR