Order Entry
Malononitrile
Malononitrile
  EHERC14733500
 :  
Malononitrile
  EHERC14733500
 :  
1 640,00 Kč
  1 * 0,25 g
  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029
 :
 :66.06 g/mol
 :13,05733
 :220 °C (1013 hPa)
 :30…32 °C
 :1.191 g/cm³ (20 °C)
 :112 °C
 :203-703-2
 :2647
 :6.1,II
 :GHS06
 :GHS09
 :109-77-3
 :C₃H₂N₂
 :MFCD00001883