Tisk…
Label printer markers for BBP3x/i3300 printers, type B-7643
Dodavatel: Brady
BRDY195556EA 11330 CZK
BRDY195556 BRDY195557 BRDY195554 BRDY195555 BRDY195561 BRDY195560 BRDY195558 BRDY195559
Label printer markers for BBP3x/i3300 printers, type B-7643
Štítky
Thermoplastic polyether polyurethane is a zero-halogen high-density, thermoplastic cable marker designed for dot matrix or thermal-transfer printing.
  • Meets the requirements of a halogen-free material per IEC 61249-2-21 (2003-11)
  • Excellent chemical, oil and water resistance
  • RoHS compliant using EU directive 2002/95/EC
  • Operating temperature of −40 to +90 °C (−40 to +193 °F)