Tisk…
Tag holder, Forkliftag kit, Scafftag®
Dodavatel: Brady
Scafftag®
BRDY138385EA 3150 CZK
BRDY138385
Tag holder, Forkliftag kit, Scafftag®
Háky na stěnu
ABS/insert - polypropylene Forkliftag holder kit.
  • Withstands 125 °C
  • Safety identification
  • With three holes
Depth of 2 mm and thickness of 0,50 mm. Preprinted in English legend.
Informace o dodávce: This includes of 10 Forkliftag holders,10 Forkliftag inserts, 1 pen.