Order Entry
Avantor® Hichrom HI-SE54 HT, GC Columns
Avantor® Hichrom HI-SE54 HT, GC Columns
Supplier:  Avantor
Certificates
undefined
Avantor® Hichrom HI-SE54 HT, GC Columns
Supplier:  Avantor
Specifications
  • Catalog No:
  • HI91-32-025-30
  • HI91-53-010-10
  • HI91-10-010-10
  • HI91-10-010-5
  • HI91-53-015-15
  • HI91-53-015-30
  • HI91-53-010-15
  • HI91-53-010-30
  • HI91-53-025-30
  • HI91-53-017-30
  • HI91-53-025-15
  • HI91-25-010-15
  • HI91-25-010-30
  • HI91-32-010-10G2
  • HI91-25-015-15
  • HI91-25-025-30
  • HI91-32-010-10
  • HI91-25-015-30
  • HI91-25-025-15
  • HI91-32-015-15
  • HI91-32-015-30
  • HI91-32-010-15
  • HI91-32-010-30
  • HI91-32-025-15
  • Phase name:
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • Phase composition:
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • Polarity:
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Crossbond:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Temperature limit:
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • USP code:
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
Specifications
Product Information
  • Catalog #
    Column Format
    Film thickness
    Ø int.
    Length
    Pk
    Availability
    Price
  • High Temperature GC Column
    High Temperature GC Column
    HI91-32-025-30
    Capillary
    0,25 µm
    0,32 mm
    30 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 21 510,00 Kč
    21 510,00 Kč
    1 * 1 KS
       21 510,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41
  • High Temperature GC Column
    High Temperature GC Column
    HI91-53-010-10
    Capillary
    0,10 µm
    0,53 mm
    10 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 16 440,00 Kč
    16 440,00 Kč
    1 * 1 KS
       16 440,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41
  • High Temperature FAST GC Column
    High Temperature FAST GC Column
    HI91-10-010-10
    Capillary
    0,10 µm
    0,10 mm
    10 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 20 390,00 Kč
    20 390,00 Kč
    1 * 1 KS
       20 390,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41
  • High Temperature FAST GC Column
    High Temperature FAST GC Column
    HI91-10-010-5
    Capillary
    0,10 µm
    0,10 mm
    5 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 17 900,00 Kč
    17 900,00 Kč
    1 * 1 KS
       17 900,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41
  • High Temperature GC Column
    High Temperature GC Column
    HI91-53-015-15
    Capillary
    0,15 µm
    0,53 mm
    15 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 17 310,00 Kč
    17 310,00 Kč
    1 * 1 KS
       17 310,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41
  • High Temperature GC Column
    High Temperature GC Column
    HI91-53-015-30
    Capillary
    0,15 µm
    0,53 mm
    30 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 25 690,00 Kč
    25 690,00 Kč
    1 * 1 KS
       25 690,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41
  • High Temperature GC Column
    High Temperature GC Column
    HI91-53-010-15
    Capillary
    0,10 µm
    0,53 mm
    15 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 17 310,00 Kč
    17 310,00 Kč
    1 * 1 KS
       17 310,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41
  • High Temperature GC Column
    High Temperature GC Column
    HI91-53-010-30
    Capillary
    0,10 µm
    0,53 mm
    30 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 25 690,00 Kč
    25 690,00 Kč
    1 * 1 KS
       25 690,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41
  • High Temperature GC Column
    High Temperature GC Column
    HI91-53-025-30
    Capillary
    0,25 µm
    0,53 mm
    30 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 25 690,00 Kč
    25 690,00 Kč
    1 * 1 KS
       25 690,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41
  • High Temperature GC Column
    High Temperature GC Column
    HI91-53-017-30
    Capillary
    0,15 µm
    0,53 mm
    30 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 25 690,00 Kč
    25 690,00 Kč
    1 * 1 KS
       25 690,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41
  • High Temperature GC Column
    High Temperature GC Column
    HI91-53-025-15
    Capillary
    0,25 µm
    0,53 mm
    15 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 17 310,00 Kč
    17 310,00 Kč
    1 * 1 KS
       17 310,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41
  • High Temperature GC Column
    High Temperature GC Column
    HI91-25-010-15
    Capillary
    0,10 µm
    0,25 mm
    15 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 16 190,00 Kč
    16 190,00 Kč
    1 * 1 KS
       16 190,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41
  • High Temperature GC Column
    High Temperature GC Column
    HI91-25-010-30
    Capillary
    0,10 µm
    0,25 mm
    30 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 21 510,00 Kč
    21 510,00 Kč
    1 * 1 KS
       21 510,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41
  • High Temperature GC column w/ integrated HI-GAP deactivated retention Gap
    High Temperature GC column w/ integrated HI-GAP deactivated retention Gap
    HI91-32-010-10G2
    Capillary
    0,10 µm
    0,32 mm
    10 m; 2 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 15 910,00 Kč
    15 910,00 Kč
    1 * 1 KS
       15 910,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41
  • High Temperature GC Column
    High Temperature GC Column
    HI91-25-015-15
    Capillary
    0,15 µm
    0,25 mm
    15 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 16 190,00 Kč
    16 190,00 Kč
    1 * 1 KS
       16 190,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41
  • High Temperature GC Column
    High Temperature GC Column
    HI91-25-025-30
    Capillary
    0,25 µm
    0,25 mm
    30 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 21 510,00 Kč
    21 510,00 Kč
    1 * 1 KS
       21 510,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41
  • High Temperature GC Column
    High Temperature GC Column
    HI91-32-010-10
    Capillary
    0,10 µm
    0,32 mm
    10 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 14 760,00 Kč
    14 760,00 Kč
    1 * 1 KS
       14 760,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41
  • High Temperature GC Column
    High Temperature GC Column
    HI91-25-015-30
    Capillary
    0,15 µm
    0,25 mm
    30 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 21 510,00 Kč
    21 510,00 Kč
    1 * 1 KS
       21 510,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41
  • High Temperature GC Column
    High Temperature GC Column
    HI91-25-025-15
    Capillary
    0,25 µm
    0,25 mm
    15 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 16 190,00 Kč
    16 190,00 Kč
    1 * 1 KS
       16 190,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41
  • High Temperature GC Column
    High Temperature GC Column
    HI91-32-015-15
    Capillary
    0,15 µm
    0,32 mm
    15 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 16 190,00 Kč
    16 190,00 Kč
    1 * 1 KS
       16 190,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41
  • High Temperature GC Column
    High Temperature GC Column
    HI91-32-015-30
    Capillary
    0,15 µm
    0,32 mm
    30 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 21 510,00 Kč
    21 510,00 Kč
    1 * 1 KS
       21 510,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41
  • High Temperature GC Column
    High Temperature GC Column
    HI91-32-010-15
    Capillary
    0,10 µm
    0,32 mm
    15 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 16 190,00 Kč
    16 190,00 Kč
    1 * 1 KS
       16 190,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41
  • High Temperature GC Column
    High Temperature GC Column
    HI91-32-010-30
    Capillary
    0,10 µm
    0,32 mm
    30 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 21 510,00 Kč
    21 510,00 Kč
    1 * 1 KS
       21 510,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41
  • High Temperature GC Column
    High Temperature GC Column
    HI91-32-025-15
    Capillary
    0,25 µm
    0,32 mm
    15 m
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 16 190,00 Kč
    16 190,00 Kč
    1 * 1 KS
       16 190,00 Kč
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Phase name
    HI-SE54 HT
    Phase composition
    5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
    Polarity
    Low-polarity stationary phase
    Crossbond
    Yes
    Temperature limit
    400 °C
    USP code
    G27, G36, G41