Order Entry
Selenium(VI) ICP Standards, (CRMs), Fluka™
Selenium(VI) ICP Standards, (CRMs), Fluka™
Selenium(VI) ICP Standards, (CRMs), Fluka™
  •  
    Element
    Symbol
    Concentration
    Matrix
    Pack type
    Pk
  • HONCE1509-100ML
    E1509-100ML
    Selenium VI
    Se
    100 mg/L
    0,1% HNO₃
    Plastic bottle
    100 mL
    1 * 100 mL
    1 * 100 mL - 3 430,00 Kč
    3 430,00 Kč
    1 * 100 mL
       3 430,00 Kč
  • HONCE1510-100ML
    E1510-100ML
    Selenium VI
    Se
    1000 mg/L
    0,1% HNO₃
    Plastic bottle
    100 mL
    1 * 100 mL
    1 * 100 mL - 4 260,00 Kč
    4 260,00 Kč
    1 * 100 mL
       4 260,00 Kč