Order Entry
Potassium AAS Standards, (CRMs), Fluka™
Potassium AAS Standards, (CRMs), Fluka™
Certificates
Potassium AAS Standards, (CRMs), Fluka™
Product Information
  • Catalog #
    Supplier #
    Element
    Symbol
    Concentration
    Matrix
    Pack type
    Pk
    Availability
    Price
  • HONCE4038-100ML
    E4038-100ML
    Potassium
    K
    1000 mg/L
    2% HNO₃
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 960,00 Kč
    960,00 Kč
    1 * 100 mL
       960,00 Kč
  • HONCE4203-500ML
    E4203-500ML
    Potassium
    K
    10000 mg/L
    12% EtOH/2% HCl
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 3 430,00 Kč
    3 430,00 Kč
    1 * 500 mL
       3 430,00 Kč
  • HONCE4038-500ML
    E4038-500ML
    Potassium
    K
    1000 mg/L
    2% HNO₃
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 1 690,00 Kč
    1 690,00 Kč
    1 * 500 mL
       1 690,00 Kč
  • HONCE4203-100ML
    E4203-100ML
    Potassium
    K
    10000 mg/L
    12% EtOH/2% HCl
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 2 310,00 Kč
    2 310,00 Kč
    1 * 100 mL
       2 310,00 Kč
  • HONCE4037-100ML
    E4037-100ML
    Potassium
    K
    1000 mg/L
    2% HCl
    Plastic bottle
    100 ml
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 960,00 Kč
    960,00 Kč
    1 * 100 mL
       960,00 Kč
  • HONCE4037-500ML
    E4037-500ML
    Potassium
    K
    1000 mg/L
    2% HCl
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 1 690,00 Kč
    1 690,00 Kč
    1 * 500 mL
       1 690,00 Kč