Tisk…
Light-duty cushion matting, Safety-Walk™ 3270E series
Dodavatel: 3M
Safety-Walk™
MMMA3270NE6EA 61280 CZK
MMMA3270NE6
Light-duty cushion matting, Safety-Walk™ 3270E series
Rohože Rohože proti únavě
Safety Walk™ Light-Duty Cushion Matting 3270E is made from continuous coiled vinyl filaments that provide comfort and support across the bottom of the shoe, contributing to a reduction in leg and back fatigue.
  • Coiled cushion matting compresses evenly across the bottom of the shoe, reducing leg and back fatigue
  • Textured top improves traction for safer footing
  • Supplied with edging to allow wheeled traffic
  • Special vinyl coils to withstand most industrial and cleaning chemicals
The textured top improves traction and allows debris and liquid to remain on the mat for easy cleanup. The mat features integral ramped edging on all sides to enable wheeled traffic movement and pedestrian safety. It is suitable for dry work areas such as assembly lines, packing areas, and security posts.