The 3M DBI-SALA® ExoFit NEX™ Harness is a safety harness built to help users withstand the harshest environments, designed using soft yet durable anti-absorbent, abrasion-resistant webbing with long-lasting strength.
- Water-repellent webbing helps keep users dry and comfortable
- Abrasion-resistant webbing for lasting strength
- Dual-lock quick connect buckles provide fast, simple and safe connection
- Integrated suspension relief straps
- Stand up dorsal D-ring
- Revolver vertical torso adjuster provides quick and easy adjustment
- Moulded lanyard keepers with breakaway holstered snap hooks
- Impact indicators make it simple to check the harness for damage and help ensure it’s safe to use
- Covered identification labels
Stay protected and comfortable when working at height with the DBI-SALA® ExoFit NEX™ Harness. This safety harness features water-repellent webbing to keep the harness dry and to reduce the attraction of mould and dirt. The abrasion-resistant webbing has lasting strength and withstands severe demands, day in and day out. The integrated suspension relief straps provide an adjustable, continuous loop for post fall suspension relief.
Revolver vertical torso adjusters provide simple and fast adjustments that eliminate loose ends and lock into place, preventing slippage. Impact indicators make it simple to check the harness for damage and help ensure it’s safe to use. Moulded lanyard keepers have easily holstered snap hooks that breakaway to avoid trip hazards. Dual-lock quick connect buckles provide lightweight one-handed use with memory-fit web-lock, helping to ensure fast, non-slip connections. Covered identification labels protect important information for the life of the harness.