Variant of concern detection kits, SNPsig®
Supplier: PRIMERDESIGN
Certificates
About this item
Specifications
- Shelf life:6 Months
- Storage temperature:–20 °C
Specifications
- ForNo. of reactionsPk
- SNPsig® EscapePLEX, PCRSNPsig® EscapePLEX, PCRPRDER00152R00152Identification of E484K, K417N, K417T and P681R variant mutations including the Alpha, Beta, Gamma and Delta Variants of Concern; plus ORF 1ab and M gene targets for confirmation of SARS-CoV-2 positive9696 Tests1 * 96 Tests: R00152: PRDER00152
- SNPsig® SARS-CoV-2 (20H/501Y.V2)SNPsig® SARS-CoV-2 (20H/501Y.V2)R00135R00135Detection of the SARS-CoV-2 Beta variant9696 Tests1 * 96 Tests: R00135: PRDER00135
- SNPsig® SARS-CoV-2 (20I/501Y.V1)SNPsig® SARS-CoV-2 (20I/501Y.V1)R00130R00130Detection of the SARS-CoV-2 Alpha variant9696 Tests1 * 96 Tests: R00130: PRDER00130
- SNPsig® SARS-CoV-2 (20J/501Y.V3)SNPsig® SARS-CoV-2 (20J/501Y.V3)R00134R00134Detection of SARS-CoV-2 Gamma variant9696 Tests1 * 96 Tests: R00134: PRDER00134
- SNPsig® SARS-CoV-2 (E484K)SNPsig® SARS-CoV-2 (E484K)R00132R00132Detection of the SARS-CoV-2 variants with the E484K mutation96961 * 96 KS: R00132: PRDER00132
- SNPsig® SARS-CoV-2 (N501Y)SNPsig® SARS-CoV-2 (N501Y)R00131R00131Detection of the SARS-CoV-2 variants with the N501Y mutation (found in all three variants of concern: Alpha, Beta and Gamma)9696 Tests1 * 96 Tests: R00131: PRDER00131
- SNPsig® SARS-CoV-2 (VUI-21APR) (Delta)SNPsig® SARS-CoV-2 (VUI-21APR) (Delta)PRDER00149R00149Detection of the Delta variant9696 Tests1 * 96 Tests: R00149: PRDER00149
- SNPsig® SARS-CoV-2 (20B/S.484K), also known as P2SNPsig® SARS-CoV-2 (20B/S.484K), also known as P2R00147R00147Detection of the SARS-CoV-2 variants with the 20B/S.484K mutation, also known as P29611 * 1 MJ: R00147: PRDER00147
- SNPsig® SARS-CoV-2 (L452R)SNPsig® SARS-CoV-2 (L452R)R00146R00146Detection of the SARS-CoV-2 variants with the L452R mutation9696 Tests1 * 96 Tests: R00146: PRDER00146
- SNPsig® VariPLEX SARS-CoV-2SNPsig® VariPLEX SARS-CoV-2R00137R00137Detects the variants originally identified: Alpha (20I/501Y.V1), Beta (20H/501Y.V2), Gamma (20J/501Y.V3) and Epsilon (20C/S:452R), and the key biologically significant mutations N501Y and E484K96961 * 96 KS: R00137: PRDER00137