Tisk…
PIG® Essentials Plus Universal mat pads
Dodavatel: New Pig
NEWPMPBH100EA 2040 CZK
NEWPMPBH100
PIG® Essentials Plus Universal mat pads
Universal Mat Pad controls spills and overspray to help keep floors dry and safe.
  • Random assembly of large-diameter meltblown poly fibres creates a lofty layer
  • Mat absorbs high volumes of liquid fast; great for catching overspray or soaking up spills
  • Ultrasonic bond points fuse layers of fibres together — Mat keeps its shape even when completely saturated
  • Top layer of lint-free SpunBond fabric provides extra strength; "dimples" add fast-wicking power
Grey colour signifies Universal sorbency for easy visual identification.