Tisk…

5-Bromo-3-indolyl β-D-glucuronide cyclohexylammonium salt

Dodavatel: Apollo Scientific

APOSBIB1407-100MGEA 4370 CZK
APOSBIB1407-100MG
5-Bromo-3-indolyl β-D-glucuronide cyclohexylammonium salt
5-Bromo-3-indolyl β-D-glucuronide cyclohexylammonium salt
Vzorec: C₁₄H₁₄BrNO₇·C₆H₁₃N
MW: 487,342 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00214410
CAS číslo: 199326-16-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR