Tisk…

3-Indolyl β-D-glucuronide cyclohexylammonium salt

Varování

APOSBIB1421-1GEA 0 CZK
APOSBIB1421-1G APOSBIB1421-500MG
3-Indolyl β-D-glucuronide cyclohexylammonium salt
3-Indolyl β-D-glucuronide cyclohexylammonium salt
Vzorec: C₁₄H₁₅NO₇·C₆H₁₃N
MW: 408,446 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00083390
CAS číslo: 216971-58-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR