Tisk…

6-Bromo-2-naphthyl α-D-galactopyranoside

Dodavatel: Apollo Scientific
Varování

APOSBIB6016-1GEA 5630 CZK
APOSBIB6016-1G APOSBIB6016-500MG
6-Bromo-2-naphthyl α-D-galactopyranoside
6-Bromo-2-naphthyl α-D-galactopyranoside
Vzorec: C₁₆H₁₇BrO₆
MW: 385,207 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066434
CAS číslo: 25997-59-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR