Tisk…

Sulpho-MBS (3-Maleimidobenzoyl-N-hydroxysulphosuccinimide ester sodium salt)

Dodavatel: Apollo Scientific
Varování

Synonyms: Sulfo MBS

APOSBICL203-100MGEA 4570 CZK
APOSBICL203-100MG
Sulpho-MBS (3-Maleimidobenzoyl-N-hydroxysulphosuccinimide ester sodium salt)
Sulpho-MBS (3-Maleimidobenzoyl-N-hydroxysulphosuccinimide ester sodium salt)

Primary amine and sulphhydryl reactive -Water soluble


Ref: Aithal,H.N,et.al (1988) J.Immunol.Meth.112, 67-70

Vzorec: C₁₅H₉N₂O₉S·Na
MW: 416,295 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01318594
CAS číslo: 92921-25-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR