N-(E-Maleimidocaproyloxy)sulphosuccinimide
Dodavatel: Apollo Scientific
|
Varování
|
APOSBICL221-50MGEA
3940
CZK
APOSBICL221-50MG
N-(E-Maleimidocaproyloxy)sulphosuccinimide
N-(E-Maleimidocaproyloxy)sulphosuccinimide
Primary amine reactive -Sulphhydryl reactive Water soluble
Ref: Kitagawa,T.et al(1981) Chem Pharm.Bull. 29, 1130Ref: Keller,O.and Rudinger, J.Helv.Chem.Acta, 58, 531
Vzorec:
C₁₄H₁₆N₂O₉S MW: 388,35 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01310272 CAS číslo: 103848-61-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...