Tisk…

3-Acetylumbelliferyl β-D-glucopyranoside ≥98%

Dodavatel: Apollo Scientific
Varování

APOSBICL4231-25MGEA 14620 CZK
APOSBICL4231-25MG APOSBICL4231-10MG APOSBICL4231-50MG
3-Acetylumbelliferyl β-D-glucopyranoside ≥98%
3-Acetylumbelliferyl β-D-glucopyranoside
Vzorec: C₁₇H₁₈O₉
MW: 366,319 g/mol
Bod tání: 238…242 °C
Storage Temperature: Freezer
MDL Number: MFCD00037378
CAS číslo: 20943-16-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR