Tisk…

Fmoc-(R)-2-(4-pentenyl)alanine

Dodavatel: Apollo Scientific
Varování

APOSBICR151-1GEA 21560 CZK
APOSBICR151-1G APOSBICR151-500MG
Fmoc-(R)-2-(4-pentenyl)alanine
Fmoc-(R)-2-(4-pentenyl)alanine
Vzorec: C₂₃H₂₅NO₄
MW: 379,449 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09879889
CAS číslo: 288617-77-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR