Tisk…

5-Bromo-6-chloro-3-indolyl β-D-glucuronide

APOSBICS0113-100MGEA 5200 CZK
APOSBICS0113-100MG APOSBICS0113-250MG APOSBICS0113-500MG
5-Bromo-6-chloro-3-indolyl β-D-glucuronide
5-Bromo-6-chloro-3-indolyl β-D-glucuronide
Storage Temperature: Ambient
MDL Number: MFCD01311740
CAS číslo: 144110-42-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR