Tisk…

Cycloartenol

Dodavatel: Apollo Scientific

APOSBIDF1002-5MGEA 25690 CZK
APOSBIDF1002-5MG
Cycloartenol
Cycloartenol
Vzorec: C₃₀H₅₀O
MW: 426,717 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00200959
CAS číslo: 469-38-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR