Tisk…

Sulphosuccinimidyl-3-(4-hydroxyphenyl)propionate sodium salt

Dodavatel: Apollo Scientific
Varování

APOSBII103-100MGEA 417 CZK
APOSBII103-100MG
Sulphosuccinimidyl-3-(4-hydroxyphenyl)propionate sodium salt
Sulphosuccinimidyl-3-(4-hydroxyphenyl)propionate sodium salt

Non-oxidative Iodination -Water soluble


Ref: Thompson, J.A. et al. (1987) Biochem. 26, 743-750Ref: Mine, K. et al. (1986) Anal.Biochem.152, 127-135

Vzorec: C₁₃H₁₂NO₈S·Na
MW: 365,291 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00078536
CAS číslo: 106827-57-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR