Tisk…

3-(N-Morpholino)propanesulphonic acid, hemisodium salt ≥98%

Synonyms: MOPS hemisodium salt3-Morpholinopropane-1-sulphonic acid, sodium salt(2:1)

APOSBIM0290-100GEA 0 CZK
APOSBIM0290-100G APOSBIM0290-1KG APOSBIM0290-500G
3-(N-Morpholino)propanesulphonic acid, hemisodium salt ≥98%
3-(N-Morpholino)propanesulphonic acid, hemisodium salt

  • Buffer


Titration > 98%
Water content (by Karl Fischer) ≥ 2.0%

Vzorec: C₁₄H₃₀N₂NaO₈S₂
MW: 441,5200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00079461
CAS číslo: 117961-20-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR