Tisk…

4-Hydroxyphenyl glyoxal (HPG)

Dodavatel: Apollo Scientific
Varování

Synonyms: HPG

APOSBIM104-100MGEA 7670 CZK
APOSBIM104-100MG
4-Hydroxyphenyl glyoxal (HPG)
4-Hydroxyphenyl glyoxal (HPG)

Modifies arginine residues


  • Protein


Ref: Yamasaki,R.B,Vega,A.and Freeney,R.E.(1980)Anal.Biochem.109, 32-40

Vzorec: C₈H₆O₃
MW: 150,131 g/mol
Bod tání: 106…112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00054984
CAS číslo: 24645-80-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR