Tisk…

Ald-Ph-PEG6-acid

Dodavatel: Apollo Scientific

APOSBIPG1018-100MGEA 20300 CZK
APOSBIPG1018-100MG
Ald-Ph-PEG6-acid
Ald-Ph-PEG6-acid
Vzorec: C₂₃H₃₅NO₁₀
MW: 485,525 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22056302
CAS číslo: 2055013-55-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR