Tisk…

m-PEG36-alcohol

APOSBIPG1573-250MGEA 22720 CZK
APOSBIPG1573-250MG
m-PEG36-alcohol
m-PEG36-alcohol
Vzorec: C₇₃H₁₄₈O₃₇
MW: 1617,934 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21363224
CAS číslo: 114740-40-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR