Tisk…

m-PEG9-amine

APOSBIPG1587-500MGEA 25530 CZK
APOSBIPG1587-500MG
m-PEG9-amine
m-PEG9-amine
Vzorec: C₁₉H₄₁NO₉
MW: 427,53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16619283
CAS číslo: 211859-73-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR