Tisk…

Reverse Triiodothyronine-[diiodophenyl-ring-13C6] hydrochloride (Reverse T3)

Dodavatel: Apollo Scientific

APOSBISC1049-1MGEA 26920 CZK
APOSBISC1049-1MG
Reverse Triiodothyronine-[diiodophenyl-ring-13C6] hydrochloride (Reverse T3)
Reverse Triiodothyronine-[diiodophenyl-ring-13C6] hydrochloride (Reverse T3)
Vzorec: [₁₃C]₆C₉H₁₂I₃NO₄·ClH
MW: 693,39 g/mol
Storage Temperature: Freezer
MDL Number: MFCD17015239
CAS číslo: 1217676-14-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR