Tisk…

2-(Fur-2-yl)-2,3,4,5-tetrahydro-1H-1,5-benzodiazepine

APOSOR01520-500MGEA 0 CZK
APOSOR01520-500MG
2-(Fur-2-yl)-2,3,4,5-tetrahydro-1H-1,5-benzodiazepine
2-(Fur-2-yl)-2,3,4,5-tetrahydro-1H-1,5-benzodiazepine
Vzorec: C₁₃H₁₄N₂O
MW: 214,263 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06245353
CAS číslo: 394655-13-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR