Tisk…

Phenylaminopyridin-3-ylacetonitrile

Dodavatel: Apollo Scientific

APOSOR01617-50MGEA 2070 CZK
APOSOR01617-50MG
Phenylaminopyridin-3-ylacetonitrile
Phenylaminopyridin-3-ylacetonitrile
Vzorec: C₁₃H₁₁N₃
MW: 209,247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06245417
CAS číslo: 904813-91-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR