2-Amino-4-methylthiophene-3-carbonitrile
Dodavatel: Apollo Scientific
|
Varování
|
APOSOR01621-1GEA
2370
CZK
APOSOR01621-1G
2-Amino-4-methylthiophene-3-carbonitrile
2-Amino-4-methylthiophene-3-carbonitrile
Vzorec:
C₆H₆N₂S MW: 138,19 g/mol Bod tání: 119…120 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00277568 CAS číslo: 4623-55-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...