Tisk…

1-(4-Aminophenyl)ethanol, N-BOC protected

APOSOR01627-50MGEA 0 CZK
APOSOR01627-50MG
1-(4-Aminophenyl)ethanol, N-BOC protected
1-(4-Aminophenyl)ethanol, N-BOC protected
Vzorec: C₁₃H₁₉NO₃
MW: 237,295 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06245420
CAS číslo: 232597-44-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR