Tisk…

2-Naphthyloxyethanamide

APOSOR10235-1GEA 0 CZK
APOSOR10235-1G
2-Naphthyloxyethanamide
2-Naphthyloxyethanamide
Vzorec: C₁₂H₁₁NO₂
MW: 201,221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234788
CAS číslo: 326887-07-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR