Tisk…

4,5-Dibromo-1-methyl-1H-imidazole 98%

Dodavatel: Apollo Scientific
Nebezpečí

APOSOR10436-5GEA 6150 CZK
APOSOR10436-5G APOSOR10436-25G APOSOR10436-1G
4,5-Dibromo-1-methyl-1H-imidazole 98%
4,5-Dibromo-1-methyl-1H-imidazole
Vzorec: C₄H₄Br₂N₂
MW: 239,896 g/mol
Bod tání: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00955568
CAS číslo: 1003-50-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR