Tisk…

Diethyl (3-aminoprop-1-yl)phosphonate oxalate 90%

Varování

APOSOR10689-250MGEA 0 CZK
APOSOR10689-250MG APOSOR10689-1G
Diethyl (3-aminoprop-1-yl)phosphonate oxalate 90%
Diethyl (3-aminoprop-1-yl)phosphonate oxalate
Storage Temperature: Ambient
MDL Number: MFCD00800257
CAS číslo: 203192-99-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR