2,5-Dibromo-4-pyridinylboronic acid
Dodavatel: Apollo Scientific
APOSOR110029-1GEA
0
CZK
APOSOR110029-1G
2,5-Dibromo-4-pyridinylboronic acid
2,5-Dibromo-4-pyridinylboronic acid
Vzorec:
C₅H₄BBr₂NO₂ MW: 280,71 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11109338 CAS číslo: 1031843-77-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...