Tisk…

2-Methyl-1,3-benzothiazole-6-sulphonamide

Dodavatel: Apollo Scientific

APOSOR110086-1GEA 8300 CZK
APOSOR110086-1G
2-Methyl-1,3-benzothiazole-6-sulphonamide
2-Methyl-1,3-benzothiazole-6-sulphonamide
Vzorec: C₈H₈N₂O₂S₂
MW: 228,291 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23381086
CAS číslo: 21431-21-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR