Methyl 2-amino-5-chloronicotinate
Dodavatel: Apollo Scientific
|
Varování
|
APOSOR110303-5GEA
3410
CZK
APOSOR110303-5G
Methyl 2-amino-5-chloronicotinate
Methyl 2-amino-5-chloronicotinate
Vzorec:
C₇H₇ClN₂O₂ MW: 186,596 g/mol Bod tání: 133…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD09031678 CAS číslo: 50735-33-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...