Tisk…

2-(3-Nitrophenyl)-1,3,4-oxadiazole

Dodavatel: Apollo Scientific

APOSOR110325-1GEA 5490 CZK
APOSOR110325-1G
2-(3-Nitrophenyl)-1,3,4-oxadiazole
2-(3-Nitrophenyl)-1,3,4-oxadiazole
Vzorec: C₈H₅N₃O₃
MW: 191,144 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00085145
CAS číslo: 5565-72-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR