Tisk…

3-Chloro-2-methyl-6-nitro-2H-indazole

Dodavatel: Apollo Scientific

APOSOR110348-1GEA 10290 CZK
APOSOR110348-1G
3-Chloro-2-methyl-6-nitro-2H-indazole
3-Chloro-2-methyl-6-nitro-2H-indazole
Vzorec: C₈H₆ClN₃O₂
MW: 211,605 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16619581
CAS číslo: 74209-40-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR